Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022494.2 | NP_071939.1 | 3 | 756 | ||
XM_005270051.4 | XP_005270108.1 | 3 | 1691 | |||
XM_006717948.3 | XP_006718011.1 | 3 | 862 | |||
XM_006717949.3 | XP_006718012.1 | 3 | 753 | |||
XM_011540091.2 | XP_011538393.1 | 3 | 744 | |||
XM_011540092.2 | XP_011538394.1 | 3 | 753 | |||
XM_011540093.1 | XP_011538395.1 | 3 | 862 | |||
XM_011540095.2 | XP_011538397.1 | 3 | 733 | |||
XM_017016561.1 | XP_016872050.1 | 3 | 682 | |||
XM_017016564.1 | XP_016872053.1 | 3 | 862 | |||
XM_017016565.1 | XP_016872054.1 | 3 | 928 | |||
XM_017016568.1 | XP_016872057.1 | 3 | 659 | |||
GenBank mRNA | AF267740.1 | - | 3 | 710 | ||
AK025605.1 | - | 3 | 711 | |||
AK223086.1 | - | 3 | 710 | |||
AK308026.1 | - | 3 | 904 | |||
BX647270.1 | - | 2 | 2184 |