Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303134.1 | NP_001290063.1 | 7 | 1274 | ||
NM_022494.2 | NP_071939.1 | 7 | 1286 | |||
XM_005270051.4 | XP_005270108.1 | 7 | 2221 | |||
XM_006717948.3 | XP_006718011.1 | 7 | 1392 | |||
XM_006717949.3 | XP_006718012.1 | 6 | 1121 | |||
XM_011540091.2 | XP_011538393.1 | 7 | 1274 | |||
XM_011540092.2 | XP_011538394.1 | 7 | 1191 | |||
XM_011540093.1 | XP_011538395.1 | 7 | 1300 | |||
XM_011540094.2 | XP_011538396.1 | 7 | 1159 | |||
XM_011540095.2 | XP_011538397.1 | 6 | 1009 | |||
XM_017016561.1 | XP_016872050.1 | 7 | 1212 | |||
XM_017016562.1 | XP_016872051.1 | 7 | 1380 | |||
XM_017016563.1 | XP_016872052.1 | 7 | 1193 | |||
XM_017016564.1 | XP_016872053.1 | 7 | 1392 | |||
XM_017016565.1 | XP_016872054.1 | 7 | 1458 | |||
XM_017016566.1 | XP_016872055.1 | 7 | 1271 | |||
XM_017016567.1 | XP_016872056.1 | 6 | 1109 | |||
XM_017016568.1 | XP_016872057.1 | 7 | 1097 | |||
XM_017016569.1 | XP_016872058.1 | 3 | 234 | |||
GenBank mRNA | AF267740.1 | - | 7 | 1240 | ||
AK025605.1 | - | 7 | 1241 | |||
AK026369.1 | - | 2 | 146 | |||
AK223086.1 | - | 7 | 1240 | |||
AK308026.1 | - | 7 | 1434 | |||
BC007213.1 | - | 7 | 1216 | 2960984 | ||
BC017434.2 | - | 7 | 1216 | 2960984 | ||
BX647270.1 | - | 6 | 2714 |