siRNA Details
Gene Symbol: CRTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042574.2 | NP_001036039.1 | 11 | 1404 | ||
NM_022769.4 | NP_073606.3 | 11 | 1404 | |||
XM_005254968.2 | XP_005255025.1 | 11 | 1372 | |||
XM_011521906.2 | XP_011520208.1 | 10 | 1276 | |||
GenBank mRNA | AK025521.1 | - | 4 | 571 | ||
AK074170.1 | - | 8 | 875 | |||
AK090443.1 | - | 11 | 1364 | |||
AK225300.1 | - | 4 | 571 | |||
AY360173.1 | - | 11 | 1225 | |||
BC074730.2 | - | 2 | 347 | |||
BC074731.2 | - | 2 | 347 | 30915456 | ||
BP238198.1 | - | 4 | 572 | |||
BP365179.1 | - | 1 | 149 | |||
DB288138.1 | - | 2 | 410 |

More Information
Panther Classification:
Molecular Function -



