Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022782.3 | NP_073619.3 | 9 | 1586 | ||
NR_103517.1 | 10 | 1650 | ||||
XM_011537740.2 | XP_011536042.1 | 9 | 1490 | |||
XM_011537741.2 | XP_011536043.1 | 9 | 1490 | |||
XM_017018673.1 | XP_016874162.1 | 9 | 1584 | |||
XM_017018674.1 | XP_016874163.1 | 9 | 1490 | |||
XM_017018676.1 | XP_016874165.1 | 9 | 1490 | |||
XM_017018677.1 | XP_016874166.1 | 9 | 1490 | |||
XM_017018679.1 | XP_016874168.1 | 10 | 1523 | |||
XM_017018680.1 | XP_016874169.1 | 8 | 1432 | |||
XM_017018681.1 | XP_016874170.1 | 8 | 1345 | |||
GenBank mRNA | AK023016.1 | - | 4 | 309 | ||
AK091181.1 | - | 5 | 915 | |||
AK096541.1 | - | 10 | 1523 | |||
BC130440.1 | - | 5 | 868 | 40146471 | ||
BC144017.1 | - | 5 | 845 |