siRNA Details
Gene Symbol: CRTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042574.2 | NP_001036039.1 | 12, 13 | 1661 | ||
NM_022769.4 | NP_073606.3 | 12, 13 | 1661 | |||
XM_005254968.2 | XP_005255025.1 | 12, 13 | 1629 | |||
XM_011521906.2 | XP_011520208.1 | 11, 12 | 1533 | |||
GenBank mRNA | AK025521.1 | - | 5, 6 | 828 | ||
AK074170.1 | - | 9, 10 | 1132 | |||
AK090443.1 | - | 12, 13 | 1621 | |||
AK225300.1 | - | 5, 6 | 828 | |||
AY360173.1 | - | 12, 13 | 1482 | |||
BC074730.2 | - | 3, 4 | 604 | |||
BC074731.2 | - | 3, 4 | 604 | 30915456 | ||
BP365179.1 | - | 2, 3 | 406 |

More Information
Panther Classification:
Molecular Function -



