Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023073.3 | NP_075561.3 | 52 | 10711 | ||
XM_005248345.3 | XP_005248402.1 | 53 | 10861 | |||
XM_005248346.3 | XP_005248403.1 | 53 | 10778 | |||
XM_005248347.3 | XP_005248404.1 | 53 | 10778 | |||
XM_005248349.3 | XP_005248406.1 | 52 | 10670 | |||
XM_005248350.3 | XP_005248407.1 | 53 | 10650 | |||
XM_006714491.3 | XP_006714554.1 | 27 | 5220 | |||
XM_011514085.2 | XP_011512387.1 | 53 | 10780 | |||
XM_011514086.2 | XP_011512388.1 | 53 | 10777 | |||
XM_011514087.2 | XP_011512389.1 | 53 | 10812 | |||
XM_011514088.2 | XP_011512390.1 | 52 | 10758 | |||
XM_011514090.2 | XP_011512392.1 | 53 | 10644 | |||
XM_011514094.2 | XP_011512396.1 | 38 | 7915 | |||
XM_017009760.1 | XP_016865249.1 | 52 | 10592 | |||
XM_017009761.1 | XP_016865250.1 | 52 | 10560 | |||
XM_017009762.1 | XP_016865251.1 | 49 | 10319 | |||
XM_017009763.1 | XP_016865252.1 | 49 | 10192 | |||
XM_017009764.1 | XP_016865253.1 | 49 | 10153 | |||
XM_017009765.1 | XP_016865254.1 | 53 | 10561 | |||
XM_017009766.1 | XP_016865255.1 | 39 | 8043 | |||
GenBank mRNA | AK023293.1 | - | 5 | 1654 | ||
AK026735.1 | - | 10 | 2141 | |||
AL833065.2 | - | 27 | 5996 |