Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTC23 SIRNA Assays ›
Gene Symbol:
TTC23
Gene Name:
tetratricopeptide repeat domain 23
Gene Aliases:
HCC-8
Chromosome Location:
Chr.15: 99136323 - 99251226 on Build GRCh38
UniGene ID:
Hs.513195
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
30520
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TTC23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001288615.1
NP_001275544.1
6
1173
NM_001288616.1
NP_001275545.1
4
564
NR_109947.1
5
834
XM_011521927.2
XP_011520229.1
2
314
XM_011521929.1
XP_011520231.1
6
881
XM_011521930.1
XP_011520232.1
5
759
XM_011521931.1
XP_011520233.1
4
902
XM_011521932.1
XP_011520234.1
5
770
XM_011521933.1
XP_011520235.1
4
703
XM_011521934.1
XP_011520236.1
6
1173
XM_011521935.1
XP_011520237.1
4
885
XM_011521936.1
XP_011520238.1
4
564
XM_011521937.1
XP_011520239.1
3
754
XM_011521939.1
XP_011520241.1
4
630
XM_011521940.2
XP_011520242.1
2
314
XM_011521941.2
XP_011520243.1
2
314
XM_011521943.2
XP_011520245.1
2
314
XM_011521944.2
XP_011520246.1
2
314
XM_011521945.2
XP_011520247.1
2
314
XM_017022509.1
XP_016877998.1
6
1332
XM_017022510.1
XP_016877999.1
5
995
XM_017022511.1
XP_016878000.1
4
769
XM_017022512.1
XP_016878001.1
4
902
XM_017022513.1
XP_016878002.1
5
770
XM_017022514.1
XP_016878003.1
4
703
XM_017022515.1
XP_016878004.1
4
885
XM_017022516.1
XP_016878005.1
4
769
XM_017022517.1
XP_016878006.1
5
686
XM_017022518.1
XP_016878007.1
4
630
GenBank mRNA
AF411456.1
-
6
1144
AK023001.1
-
2
132
AK023230.1
-
6
872
AK291690.1
-
4
561
BC015728.1
-
5
733
4857155
DA743180.1
-
3
363
DB088037.1
-
3
498
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-bn9m5:80/10.239.167.51:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0