Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUEDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024040.2 | NP_076945.2 | 3 | 306 | ||
GenBank mRNA | AB025425.1 | - | 2 | 155 | ||
AB232358.1 | - | 2 | 155 | |||
BC000262.1 | - | 3 | 236 | 3353174 | ||
BM756951.1 | - | 3 | 306 | |||
CX165866.1 | - | 3 | 190 |