Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC25A23 SIRNA Assays ›
          Gene Symbol:
          SLC25A23
          Gene Name:
          solute carrier family 25 member 23
          Gene Aliases:
          APC2, MCSC2, SCaMC-3
          Chromosome Location:
          Chr.19: 6436081 - 6460221 on Build GRCh38
          UniGene ID:
          Hs.356231
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 30813

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC25A23
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_024103.2 NP_077008.2 2, 3 429
          XM_011528274.1 XP_011526576.1 2, 3 880
          XM_011528275.1 XP_011526577.1 2, 3 880
          XM_011528276.1 XP_011526578.1 2, 3 880
          XM_011528277.1 XP_011526579.1 2, 3 880
          XM_011528278.1 XP_011526580.1 2, 3 880
          XM_011528279.1 XP_011526581.1 2, 3 880
          XM_011528280.1 XP_011526582.1 2, 3 880
          XM_011528281.1 XP_011526583.1 2, 3 880
          XM_011528282.1 XP_011526584.1 2, 3 880
          XM_011528283.1 XP_011526585.1 2, 3 880
          XM_011528284.1 XP_011526586.1 2, 3 880
          XM_011528285.1 XP_011526587.1 2, 3 880
          XM_017027285.1 XP_016882774.1 2, 3 880
          XM_017027286.1 XP_016882775.1 2, 3 880
          XM_017027287.1 XP_016882776.1 2, 3 880
          XM_017027288.1 XP_016882777.1 2, 3 880
          XM_017027289.1 XP_016882778.1 2, 3 880
          XM_017027290.1 XP_016882779.1 2, 3 880
          XM_017027291.1 XP_016882780.1 2, 3 880
          XM_017027292.1 XP_016882781.1 2, 3 880
          GenBank mRNA AJ512835.1 - 2, 3 394
          AJ619962.1 - 2, 3 384
          AJ619988.1 - 2, 3 394
          AK054901.1 - 1, 2 126
          AK294514.1 - 2, 3 287
          AY750170.1 - 2, 3 394
          BC001656.1 - 2, 3 410 3356718
          BG706035.1 - 2, 3 456

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          mitochondrial carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of oxidative phosphorylation
          translation
          mitochondrial calcium ion transport
          calcium ion transmembrane import into mitochondrion
          regulation of cellular respiration
          regulation of sequestering of calcium ion
          adenine nucleotide transport
          cellular response to calcium ion
          urea homeostasis
          structural constituent of ribosome
          calcium ion binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gl4ql:80/100.66.76.174:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0