Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024112.3 | NP_077017.1 | 2 | 683 | ||
XM_011519004.1 | XP_011517306.1 | 2 | 631 | |||
GenBank mRNA | AK022885.1 | - | 1 | 2832 | ||
BC001857.2 | - | 2 | 587 | 3529116 | ||
BC008887.2 | - | 2 | 587 | 3027807 | ||
CN360246.1 | - | 2 | 483 |