Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317929.1 | NP_001304858.1 | 3 | 1102 | ||
NM_024330.1 | NP_077306.1 | 3 | 1102 | |||
GenBank mRNA | AJ577572.1 | - | 3 | 1395 | ||
AK027499.1 | - | 3 | 1006 | |||
AK074134.1 | - | 2 | 1428 | |||
AK075377.1 | - | 3 | 1095 | |||
AK123997.1 | - | 4 | 566 | |||
AK125102.1 | - | 3 | 2040 | |||
AY358409.1 | - | 3 | 1090 | |||
BC003041.1 | - | 3 | 1102 | 2822875 | ||
BC003654.2 | - | 2 | 151 | |||
BC009916.2 | - | 3 | 1102 | 2822875 | ||
BC029792.1 | - | 3 | 970 | 5195359 |