Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FASTKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024091.3 | NP_076996.2 | 2, 3 | 1618 | ||
XM_006714498.1 | XP_006714561.1 | 2, 3 | 1529 | |||
GenBank mRNA | AK026927.1 | - | 2, 3 | 1583 | ||
BC001295.1 | - | 1, 2 | 1110 | |||
BC113563.1 | - | 1, 2 | 1533 | 8322615 | ||
BC143517.1 | - | 1, 2 | 1533 | |||
BC143518.1 | - | 2, 3 | 970 |