Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP13A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024524.3 | NP_078800.3 | 10, 11 | 1380 | ||
XM_005269357.3 | XP_005269414.1 | 10, 11 | 1401 | |||
XM_011513120.2 | XP_011511422.1 | 11, 12 | 1502 | |||
XM_011513121.1 | XP_011511423.1 | 10, 11 | 1380 | |||
XM_011513122.1 | XP_011511424.1 | 10, 11 | 1074 | |||
XM_011513123.2 | XP_011511425.1 | 11, 12 | 1507 | |||
XM_011513124.2 | XP_011511426.1 | 11, 12 | 1507 | |||
GenBank mRNA | AJ306929.1 | - | 1, 2 | 1904 | ||
BP313821.1 | - | 4, 5 | 427 | |||
DR005713.1 | - | 3, 4 | 261 |