Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP70 SIRNA Assays ›
Gene Symbol:
CEP70
Gene Name:
centrosomal protein 70
Gene Aliases:
BITE
Chromosome Location:
Chr.3: 138494339 - 138594383 on Build GRCh38
UniGene ID:
Hs.531962
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31055
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP70
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001288964.1
NP_001275893.1
5
478
NM_001288965.1
NP_001275894.1
4
422
NM_001288966.1
NP_001275895.1
5
496
NM_001320598.1
NP_001307527.1
5
548
NM_001320599.1
NP_001307528.1
5
496
NM_001320600.1
NP_001307529.1
4
362
NM_024491.3
NP_077817.2
5
496
XM_005247805.3
XP_005247862.1
3
216
XM_011513209.2
XP_011511511.1
3
216
XM_017007270.1
XP_016862759.1
3
216
XM_017007271.1
XP_016862760.1
5
398
XM_017007272.1
XP_016862761.1
5
736
XM_017007273.1
XP_016862762.1
4
341
XM_017007274.1
XP_016862763.1
4
761
XM_017007275.1
XP_016862764.1
5
832
XM_017007278.1
XP_016862767.1
5
468
XM_017007279.1
XP_016862768.1
4
344
XM_017007280.1
XP_016862769.1
4
341
XM_017007282.1
XP_016862771.1
5
467
XM_017007283.1
XP_016862772.1
5
398
GenBank mRNA
AF202146.1
-
5
350
AF289495.1
-
5
350
AK023098.1
-
3
211
AK225440.1
-
5
361
AK295866.1
-
5
367
AK299003.1
-
4
287
AK301913.1
-
5
355
AK314314.1
-
5
468
BC016050.2
-
4
363
BC030598.2
-
5
398
4829481
BM908227.1
-
358
BX331994.2
-
5
505
DC305447.1
-
5
500
DC383928.1
-
5
534
DN993112.1
-
5
490
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-rxbn5:80/10.239.162.255:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0