Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTDC1 SIRNA Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
Hmat-Xa, mat-Xa
Chromosome Location:
Chr.2: 143945787 - 144333092 on Build GRCh38
UniGene ID:
Hs.44780
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31273
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GTDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006636.3
NP_001006637.1
7
1013
NM_001164629.3
NP_001158101.1
7
998
NM_001284233.1
NP_001271162.1
5
822
NM_001284234.1
NP_001271163.1
5
713
NM_001284235.1
NP_001271164.1
7
980
NM_001284238.1
NP_001271167.1
9
1212
NM_024659.4
NP_078935.2
7
980
XM_005263774.2
XP_005263831.1
8
1097
XM_005263782.2
XP_005263839.1
8
1113
XM_005263784.2
XP_005263841.1
6
972
XM_011511843.2
XP_011510145.1
8
1146
XM_011511855.2
XP_011510157.1
6
1090
XM_011511856.2
XP_011510158.1
6
872
XM_017004917.1
XP_016860406.1
12
1768
XM_017004918.1
XP_016860407.1
10
1360
XM_017004919.1
XP_016860408.1
8
1246
XM_017004920.1
XP_016860409.1
10
4402
XM_017004921.1
XP_016860410.1
9
1195
XM_017004922.1
XP_016860411.1
9
1372
XM_017004923.1
XP_016860412.1
8
1160
XM_017004924.1
XP_016860413.1
10
1356
XM_017004925.1
XP_016860414.1
7
1018
XM_017004926.1
XP_016860415.1
5
949
XM_017004927.1
XP_016860416.1
7
980
XM_017004928.1
XP_016860417.1
9
1213
XM_017004929.1
XP_016860418.1
7
950
XM_017004930.1
XP_016860419.1
7
1601
XM_017004931.1
XP_016860420.1
7
1014
XM_017004932.1
XP_016860421.1
7
984
XM_017004933.1
XP_016860422.1
7
966
XM_017004934.1
XP_016860423.1
8
4173
XM_017004935.1
XP_016860424.1
9
2043
XM_017004936.1
XP_016860425.1
8
1220
XM_017004937.1
XP_016860426.1
6
1101
XM_017004938.1
XP_016860427.1
6
1099
XM_017004939.1
XP_016860428.1
8
1149
XM_017004940.1
XP_016860429.1
5
950
XM_017004941.1
XP_016860430.1
5
949
XM_017004942.1
XP_016860431.1
7
996
XM_017004943.1
XP_016860432.1
7
1662
XM_017004944.1
XP_016860433.1
7
995
XM_017004945.1
XP_016860434.1
9
1213
XM_017004946.1
XP_016860435.1
7
950
XM_017004947.1
XP_016860436.1
7
1633
XM_017004948.1
XP_016860437.1
8
4173
GenBank mRNA
AB201269.1
-
4
734
AB597184.1
-
4
734
AK021815.1
-
1
189
AK091684.1
-
5
826
AK291357.1
-
7
928
AY281366.1
-
7
1013
BC017741.2
-
8
1020
4390876
BC061699.1
-
7
920
4608518
BX431825.2
-
1
136
BX647436.1
-
5
695
BX647773.1
-
9
1151
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
transferase activity, transferring glycosyl groups
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline