Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEF2A SIRNA Assays ›
          Gene Symbol:
          MEF2A
          Gene Name:
          myocyte enhancer factor 2A
          Gene Aliases:
          ADCAD1, RSRFC4, RSRFC9, mef2
          Chromosome Location:
          Chr.15: 99565455 - 99716488 on Build GRCh38
          UniGene ID:
          Hs.268675
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 3140

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MEF2A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005587.3 NP_005578.2 4, 5 667
          XM_005254914.2 XP_005254971.1 4, 5 632
          XM_005254916.3 XP_005254973.1 4, 5 632
          XM_011521571.2 XP_011519873.1 4, 5 632
          XM_011521572.2 XP_011519874.1 4, 5 577
          XM_011521573.2 XP_011519875.1 4, 5 607
          XM_011521576.2 XP_011519878.1 3, 4 521
          XM_011521577.2 XP_011519879.1 3, 4 455
          XM_011521578.2 XP_011519880.1 3, 4 1114
          XM_011521579.2 XP_011519881.1 3, 4 531
          XM_011521581.2 XP_011519883.1 4, 5 895
          XM_011521582.2 XP_011519884.1 5, 6 1100
          XM_011521590.2 XP_011519892.1 4, 5 632
          XM_017022190.1 XP_016877679.1 3, 4 521
          XM_017022191.1 XP_016877680.1 3, 4 513
          XM_017022192.1 XP_016877681.1 3, 4 510
          XM_017022193.1 XP_016877682.1 4, 5 705
          XM_017022194.1 XP_016877683.1 3, 4 521
          XM_017022203.1 XP_016877692.1 4, 5 895
          GenBank mRNA BC013437.2 - 4, 5 585 3896491
          KU178181.1 - 2, 3 266
          KU178182.1 - 2, 3 266
          X63381.1 - 3, 4 494
          X68505.1 - 4, 5 680

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001014035, XM_008759542, XM_008759540, GU646868, BC081907.
          This siRNA also targets the following transcripts in Mouse: NM_001291191, NM_001291192, BC096598, AK153751.

          Panther Classification:

          Molecular Function -

          MADS box transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial genome maintenance
          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          apoptotic process
          heart development
          muscle organ development
          positive regulation of cardiac muscle hypertrophy
          positive regulation of transcription from RNA polymerase II promoter
          mitochondrion distribution
          dendrite morphogenesis
          positive regulation of muscle cell differentiation
          ventricular cardiac myofibril assembly
          cardiac conduction
          ERK5 cascade
          cellular response to calcium ion
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          RNA polymerase II transcription coactivator activity
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          protein kinase binding
          activating transcription factor binding
          histone acetyltransferase binding
          histone deacetylase binding
          sequence-specific DNA binding
          SMAD binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0