Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281476.2 | NP_001268405.1 | 11 | 2411 | ||
NM_001322046.1 | NP_001308975.1 | 12 | 3213 | |||
NM_001322048.1 | NP_001308977.1 | 11 | 2468 | |||
NM_001322049.1 | NP_001308978.1 | 10 | 2342 | |||
NM_024622.5 | NP_078898.3 | 12 | 2540 | |||
NR_104020.2 | 11 | 2411 | ||||
XM_006712750.3 | XP_006712813.1 | 11 | 2466 | |||
XM_006712751.3 | XP_006712814.1 | 8 | 2551 | |||
XM_017004911.1 | XP_016860400.1 | 12 | 3853 | |||
XM_017004912.1 | XP_016860401.1 | 7 | 2422 | |||
GenBank mRNA | AB058703.1 | - | 10 | 1722 | ||
AK025554.1 | - | 9 | 1652 | |||
AK055892.1 | - | 11 | 2322 | |||
AK074302.1 | - | 12 | 2431 | |||
AK130797.1 | - | 11 | 2335 | |||
AK225293.1 | - | 9 | 1652 | |||
AL832058.1 | - | 12 | 2423 | |||
BC032687.2 | - | 11 | 1869 | 5581443 |