Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSPP1 SIRNA Assays ›
Gene Symbol:
CSPP1
Gene Name:
centrosome and spindle pole associated protein 1
Gene Aliases:
CSPP, JBTS21
Chromosome Location:
Chr.8: 67062417 - 67196614 on Build GRCh38
UniGene ID:
Hs.370147
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31671
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSPP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291339.1
NP_001278268.1
19, 20
2852
NM_024790.6
NP_079066.5
22, 23
2852
XM_005251305.3
XP_005251362.2
25, 26
3336
XM_006716474.2
XP_006716537.2
24, 25
3183
XM_006716477.2
XP_006716540.2
20, 21
2805
XM_011517598.1
XP_011515900.1
26, 27
3378
XM_011517599.1
XP_011515901.1
25, 26
3354
XM_011517600.1
XP_011515902.1
24, 25
3312
XM_011517601.1
XP_011515903.1
24, 25
3273
XM_011517602.1
XP_011515904.1
23, 24
3231
XM_011517603.1
XP_011515905.1
25, 26
3061
XM_011517607.1
XP_011515909.1
24, 25
2941
XM_011517608.1
XP_011515910.1
23, 24
2938
XM_011517609.1
XP_011515911.1
22, 23
3060
XM_011517611.2
XP_011515913.1
10, 11
1437
XM_017013847.1
XP_016869336.1
24, 25
3237
XM_017013848.1
XP_016869337.1
23, 24
3213
XM_017013849.1
XP_016869338.1
23, 24
3174
XM_017013850.1
XP_016869339.1
22, 23
3132
XM_017013851.1
XP_016869340.1
22, 23
2896
XM_017013852.1
XP_016869341.1
21, 22
2979
XM_017013853.1
XP_016869342.1
21, 22
2743
XM_017013854.1
XP_016869343.1
19, 20
2781
XM_017013855.1
XP_016869344.1
17, 18
2547
XM_017013856.1
XP_016869345.1
17, 18
2365
XM_017013857.1
XP_016869346.1
18, 19
2415
XM_017013858.1
XP_016869347.1
16, 17
2163
XM_017013859.1
XP_016869348.1
12, 13
1565
GenBank mRNA
AJ583433.1
-
19, 20
2852
AK026143.1
-
299
AK308372.1
-
6, 7
828
AM156947.1
-
22, 23
2853
BC022867.1
-
4, 5
351
BX649147.1
-
22, 23
2854
Back To Top
More Information
Panther Classification:
Molecular Function -
centromere DNA-binding protein
Gene Ontology Categories:
Function(s)
positive regulation of cytokinesis
positive regulation of cell division
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0