Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198.3 | NP_001189.2 | 2 | 496 | ||
XM_006715550.3 | XP_006715613.1 | 2 | 1474 | |||
XM_011536062.2 | XP_011534364.1 | 2 | 1470 | |||
XM_011536063.2 | XP_011534365.1 | 2 | 548 | |||
XM_017011187.1 | XP_016866676.1 | 2 | 283 | |||
GenBank mRNA | AF084199.1 | - | 2 | 377 | ||
AK289556.1 | - | 2 | 440 | |||
AL832963.1 | - | 2 | 430 | |||
AY198414.1 | - | 2 | 496 |