Search
Search

MCMBP| Gene Symbol: MCMBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256378.1 | NP_001243307.1 | 7, 8 | 1033 | ||
| NM_001256379.1 | NP_001243308.1 | 7, 8 | 843 | |||
| NM_024834.3 | NP_079110.1 | 7, 8 | 1033 | |||
| XM_005270157.4 | XP_005270214.1 | 7, 8 | 1115 | |||
| XM_011540170.1 | XP_011538472.1 | 7, 8 | 874 | |||
| XM_017016663.1 | XP_016872152.1 | 7, 8 | 1115 | |||
| XM_017016664.1 | XP_016872153.1 | 7, 8 | 874 | |||
| GenBank mRNA | AK001928.1 | - | 3, 4 | 1205 | ||
| AK023143.1 | - | 7, 8 | 904 | |||
| AK094075.1 | - | 7, 8 | 837 | |||
| BC004183.2 | - | 7, 8 | 999 | 2961492 | ||
| BC007219.1 | - | 7, 8 | 999 | 2961492 | ||
| CR457299.1 | - | 7, 8 | 734 | |||
| KU178788.1 | - | 7, 8 | 734 | |||
| KU178789.1 | - | 7, 8 | 703 | |||