Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBSCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024186.1 | - | 1 | 798 | ||
BC098118.1 | - | 1 | 360 | |||
BC098262.1 | - | 1 | 360 | |||
BC098292.1 | - | 1 | 360 | |||
BC099731.1 | - | 1 | 360 | |||
BC114382.1 | - | 1 | 360 | |||
BC114461.1 | - | 1 | 360 |