Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024939.2 | NP_079215.2 | 4, 5 | 751 | ||
XM_005256153.4 | XP_005256210.1 | 4, 5 | 693 | |||
XM_006721273.3 | XP_006721336.1 | 4, 5 | 759 | |||
XM_011523338.2 | XP_011521640.1 | 3, 4 | 651 | |||
GenBank mRNA | AK025571.1 | - | 4, 5 | 650 | ||
BC030146.2 | - | 4, 5 | 573 | 4810948 | ||
BC069241.1 | - | 4, 5 | 1102 | |||
BU943912.1 | - | 4, 5 | 585 | |||
LN680438.1 | - | 4, 5 | 563 | |||
LN680439.1 | - | 4, 5 | 563 |