Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144875.1 | NP_001138347.1 | 4 | 651 | ||
NM_001144876.1 | NP_001138348.1 | 3 | 427 | |||
NM_001308235.1 | NP_001295164.1 | 4 | 843 | |||
NM_001308236.1 | NP_001295165.1 | 4 | 666 | |||
NM_024872.2 | NP_079148.2 | 4 | 622 | |||
XM_005265985.3 | XP_005266042.1 | 4 | 679 | |||
XM_005265987.1 | XP_005266044.1 | 4 | 679 | |||
XM_011534654.2 | XP_011532956.1 | 4 | 556 | |||
XM_011534655.2 | XP_011532957.1 | 4 | 732 | |||
XM_011534656.2 | XP_011532958.1 | 4 | 533 | |||
XM_011534657.2 | XP_011532959.1 | 3 | 383 | |||
XM_017009883.1 | XP_016865372.1 | 2 | 354 | |||
XM_017009884.1 | XP_016865373.1 | 2 | 354 | |||
GenBank mRNA | AK026223.1 | - | 4 | 617 | ||
AK097258.1 | - | 3 | 383 | |||
AL583057.3 | - | 4 | 795 | |||
BC004564.2 | - | 4 | 610 | 4054381 | ||
BC004867.2 | - | 2 | 718 | |||
BI489608.1 | - | 4 | 644 |