Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEBPA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026887.1 | 1 | 582 | |||
GenBank mRNA | AK022417.1 | - | 1 | 484 | ||
AL522369.3 | - | 1 | 582 | |||
BC027959.1 | - | 1 | 568 | |||
BC113872.1 | - | 1 | 369 | |||
BC114482.1 | - | 1 | 369 |