Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWH43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286791.1 | NP_001273720.1 | 3 | 364 | ||
NM_025087.2 | NP_079363.2 | 3 | 445 | |||
XM_011513755.1 | XP_011512057.1 | 3 | 446 | |||
XM_011513756.2 | XP_011512058.1 | 3 | 477 | |||
XM_011513757.2 | XP_011512059.1 | 3 | 503 | |||
XM_011513758.1 | XP_011512060.1 | 3 | 446 | |||
GenBank mRNA | AK025164.1 | - | 3 | 445 | ||
AK300495.1 | - | 3 | 367 | |||
AK316307.1 | - | 3 | 337 | |||
BC137387.1 | - | 3 | 428 | 9021385 | ||
BC137388.1 | - | 3 | 428 | 9021386 | ||
DA426878.1 | - | 3 | 445 |