Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534472.2 | XP_011532774.1 | 1 | 315 | ||
GenBank mRNA | AK025130.1 | - | 1 | 127 |