Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160227.1 | NP_001153699.1 | 37 | 6775 | ||
NM_025137.3 | NP_079413.3 | 39 | 7114 | |||
XM_006720700.1 | XP_006720763.1 | 39 | 6970 | |||
XM_017022634.1 | XP_016878123.1 | 38 | 7006 | |||
XM_017022636.1 | XP_016878125.1 | 28 | 4923 | |||
GenBank mRNA | AB058743.2 | - | 39 | 7084 | ||
AB470309.1 | - | 39 | 7083 | |||
AB470310.1 | - | 39 | 7083 | |||
AK025092.1 | - | 10 | 1235 | |||
AK057869.1 | - | 7 | 1454 | |||
AK225186.1 | - | 10 | 1235 | |||
BC024161.1 | - | 9 | 1200 | |||
BC067798.1 | - | 10 | 1476 | |||
BC094704.1 | - | 10 | 1608 | |||
BC150640.1 | - | 37 | 6772 | 9057010 | ||
BC153879.1 | - | 39 | 7084 | 8860108 |