siRNA Details
Gene Symbol: HM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030789.3 | NP_110416.1 | 5, 6 | 686 | ||
NM_178580.2 | NP_848695.1 | 5, 6 | 686 | |||
NM_178581.2 | NP_848696.1 | 5, 6 | 686 | |||
GenBank mRNA | AF172086.1 | - | 5, 6 | 637 | ||
AF483215.1 | - | 5, 6 | 632 | |||
AF515663.1 | - | 5, 6 | 544 | |||
AJ345029.1 | - | 5, 6 | 629 | |||
AJ420895.1 | - | 5, 6 | 568 | |||
AK075283.1 | - | 5, 6 | 646 | |||
AK314410.1 | - | 5, 6 | 654 | |||
AY169310.1 | - | 5, 6 | 643 | |||
AY169311.1 | - | 5, 6 | 643 | |||
AY169312.1 | - | 5, 6 | 643 | |||
BC008938.2 | - | 5, 6 | 657 | 2961249 | ||
BC008959.2 | - | 5, 6 | 657 | 2961249 | ||
BC062595.1 | - | 5, 6 | 637 | 5245982 | ||
BI552780.1 | - | 5, 6 | 672 | |||
DQ168450.1 | - | 5, 6 | 638 | |||
KU178790.1 | - | 5, 6 | 544 | |||
KU178791.1 | - | 5, 6 | 544 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
