Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317950.1 | NP_001304879.1 | 17 | 3658 | ||
NM_001317952.1 | NP_001304881.1 | 16 | 3216 | |||
NM_030767.5 | NP_110394.3 | 17 | 3603 | |||
XM_005252244.2 | XP_005252301.1 | 19 | 3775 | |||
XM_005252245.1 | XP_005252302.1 | 18 | 3633 | |||
XM_005252247.4 | XP_005252304.1 | 17 | 3654 | |||
XM_005252248.2 | XP_005252305.1 | 17 | 3771 | |||
XM_006717294.1 | XP_006717357.1 | 17 | 3587 | |||
XM_011519063.2 | XP_011517365.1 | 16 | 3266 | |||
XM_011519064.2 | XP_011517366.1 | 17 | 3771 | |||
XM_011519065.2 | XP_011517367.2 | 17 | 3246 | |||
XM_017015172.1 | XP_016870661.1 | 18 | 3373 | |||
GenBank mRNA | AF286341.1 | - | 7 | 1158 | ||
AK024431.1 | - | 15 | 2551 | |||
AK074040.1 | - | 16 | 2646 | |||
AK131082.2 | - | 17 | 3658 | |||
AK160382.1 | - | 17 | 3660 | |||
AL713755.1 | - | 1 | 33 | |||
AL833220.1 | - | 14 | 2080 | |||
AY703039.1 | - | 18 | 3258 | |||
AY703040.1 | - | 16 | 3216 | |||
AY703043.1 | - | 16 | 3576 | |||
AY703044.1 | - | 15 | 3270 | |||
AY703045.1 | - | 8 | 1353 | |||
AY703046.1 | - | 4 | 479 | |||
BC042202.1 | - | 3 | 333 | |||
BC055285.1 | - | 17 | 3603 | 6143155 |