Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EIF2AK2 SIRNA Assays ›
          Gene Symbol:
          EIF2AK2
          Gene Name:
          eukaryotic translation initiation factor 2 alpha kinase 2
          Gene Aliases:
          EIF2AK1, PKR, PPP1R83, PRKR
          Chromosome Location:
          Chr.2: 37099210 - 37157065 on Build GRCh38
          UniGene ID:
          Hs.131431
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 328

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EIF2AK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135651.2 NP_001129123.1 10, 11 1116
          NM_002759.3 NP_002750.1 10, 11 1350
          XM_011532987.2 XP_011531289.1 10, 11 1050
          XM_017004503.1 XP_016859992.1 10, 11 1046
          GenBank mRNA AK290655.1 - 10, 11 1036
          AK313818.1 - 9, 10 873
          BC007769.2 - 9, 10 892
          BC022314.1 - 10, 11 1004
          BC040851.1 - 10, 11 1116
          BC057805.1 - 10, 11 1021
          BC093676.1 - 8, 9 821 7939521
          BC101475.1 - 8, 9 821 8068981
          BC143228.1 - 8, 9 820
          BC143230.1 - 8, 9 821 9051731
          M35663.1 - 9, 10 979
          M85294.1 - 8, 9 823

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          positive regulation of cytokine production
          transcription, DNA-templated
          translation
          protein phosphorylation
          negative regulation of cell proliferation
          response to mechanical stimulus
          response to virus
          response to toxic substance
          regulation of translational initiation by eIF2 alpha phosphorylation
          negative regulation of translation
          peptidyl-tyrosine phosphorylation
          evasion or tolerance by virus of host immune response
          endoplasmic reticulum unfolded protein response
          response to lipopolysaccharide
          positive regulation of chemokine production
          positive regulation of stress-activated MAPK cascade
          response to vitamin E
          negative regulation of osteoblast proliferation
          regulation of protein phosphatase type 2A activity
          cellular response to amino acid starvation
          response to interferon-alpha
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          response to exogenous dsRNA
          negative regulation of viral genome replication
          innate immune response
          protein autophosphorylation
          positive regulation of NF-kappaB transcription factor activity
          defense response to virus
          regulation of NLRP3 inflammasome complex assembly
          positive regulation of NIK/NF-kappaB signaling
          regulation of hematopoietic progenitor cell differentiation
          regulation of hematopoietic stem cell proliferation
          regulation of hematopoietic stem cell differentiation
          double-stranded RNA binding
          protein kinase activity
          protein serine/threonine kinase activity
          eukaryotic translation initiation factor 2alpha kinase activity
          non-membrane spanning protein tyrosine kinase activity
          protein binding
          ATP binding
          protein phosphatase type 2A regulator activity
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-cdcg2:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0