Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037333.2 | NP_001032410.1 | 31 | 5771 | ||
NM_001291721.1 | NP_001278650.1 | 30 | 5693 | |||
NM_001291722.1 | NP_001278651.1 | 32 | 5846 | |||
NM_014376.3 | NP_055191.2 | 31 | 5897 | |||
GenBank mRNA | AF132197.1 | - | 1 | 907 | ||
AK172724.1 | - | 1 | 1662 | |||
BC026892.2 | - | 1 | 593 | |||
BC066933.1 | - | 1 | 1094 |