Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030789.3 | NP_110416.1 | 12 | 1191 | ||
NM_178580.2 | NP_848695.1 | 13 | 1394 | |||
NM_178581.2 | NP_848696.1 | 13 | 1338 | |||
GenBank mRNA | AF172086.1 | - | 12 | 1139 | ||
AF483215.1 | - | 12 | 1137 | |||
AF515663.1 | - | 12 | 1049 | |||
AJ345029.1 | - | 12 | 1134 | |||
AJ420895.1 | - | 12 | 1073 | |||
AK075283.1 | - | 12 | 1151 | |||
AK314410.1 | - | 12 | 1159 | |||
AY169310.1 | - | 13 | 1351 | |||
AY169311.1 | - | 12 | 1148 | |||
AY169312.1 | - | 13 | 1295 | |||
BC008938.2 | - | 12 | 1162 | 2961249 | ||
BC008959.2 | - | 12 | 1162 | 2961249 | ||
BC062595.1 | - | 12 | 1142 | 5245982 | ||
DQ168450.1 | - | 13 | 1346 | |||
KU178790.1 | - | 12 | 1049 | |||
KU178791.1 | - | 13 | 1150 |