Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TLR10 SIRNA Assays ›
          Gene Symbol:
          TLR10
          Gene Name:
          toll like receptor 10
          Gene Aliases:
          CD290
          Chromosome Location:
          Chr.4: 38772238 - 38782990 on Build GRCh38
          UniGene ID:
          Hs.120551
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 32946

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TLR10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001017388.2 NP_001017388.1 2 223
          NM_001195106.1 NP_001182035.1 3 540
          NM_001195107.1 NP_001182036.1 3 412
          NM_001195108.1 NP_001182037.1 3 443
          NM_030956.3 NP_112218.2 4 729
          XM_011513760.2 XP_011512062.1 2 117
          XM_011513761.2 XP_011512063.1 5 810
          XM_011513762.2 XP_011512064.1 4 490
          GenBank mRNA AB445680.1 - 1 91
          AF296673.1 - 3 577
          AK292026.1 - 2 209
          AK313468.1 - 2 360
          AY358300.1 - 2 201
          BC089406.1 - 2 575 3066588
          BC109111.2 - 1 698 40035401
          BC109112.2 - 2 241 40035404
          BP383038.1 - 3 442
          DA312129.1 - 3 408
          DA643313.1 - 2 223
          DQ011502.1 - 1 91
          DQ011503.1 - 1 91
          DQ011504.1 - 1 91
          DQ011505.1 - 1 91
          DQ011506.1 - 1 91
          DQ011507.1 - 1 91
          DQ011508.1 - 1 91
          DQ011509.1 - 1 91
          DQ011510.1 - 1 91
          DQ011511.1 - 1 91
          DQ011512.1 - 1 91
          DQ011513.1 - 1 91
          DQ011514.1 - 1 91
          DQ011515.1 - 1 91
          DQ011516.1 - 1 91

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          toll-like receptor signaling pathway
          MyD88-dependent toll-like receptor signaling pathway
          inflammatory response
          immune response
          toll-like receptor 10 signaling pathway
          innate immune response
          regulation of cytokine secretion
          positive regulation of inflammatory response
          transmembrane signaling receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0