Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201366.1 | NP_001188295.1 | 6 | 437 | ||
NM_031427.3 | NP_113615.2 | 5 | 391 | |||
XM_017021679.1 | XP_016877168.1 | 6 | 425 | |||
GenBank mRNA | AF542071.1 | - | 5 | 267 | ||
AI242197.1 | - | 3 | 242 | |||
AK026653.1 | - | 2 | 49 | |||
AK315392.1 | - | 5 | 286 | |||
AL833654.1 | - | 4 | 147 | |||
BC005343.1 | - | 5 | 267 | 3930213 |