Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135635.1 | NP_001129107.1 | 2 | 673 | ||
NM_031450.3 | NP_113638.2 | 2 | 670 | |||
GenBank mRNA | AF073483.1 | - | 2 | 579 | ||
BC004414.1 | - | 1 | 497 | |||
BC010512.1 | - | 1 | 664 | 4150027 | ||
BC013185.1 | - | 2 | 670 | 4346483 | ||
BM705907.1 | - | 1 | 533 | |||
CX866110.1 | - | 2 | 649 |