Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TWIST1 SIRNA Assays ›
Gene Symbol:
TWIST1
Gene Name:
twist family bHLH transcription factor 1
Gene Aliases:
ACS3, BPES2, BPES3, CRS, CRS1, CSO, SCS, TWIST, bHLHa38
Chromosome Location:
Chr.7: 19113047 - 19117672 on Build GRCh38
UniGene ID:
Hs.644998
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
3353
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TWIST1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000474.3
NP_000465.1
2
1031
GenBank mRNA
BC036704.2
-
2
1031
4125830
X99268.1
-
2
801
Y11178.1
-
2
115
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011658, BC083139, BC033434, M63650.
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
ossification
osteoblast differentiation
in utero embryonic development
neuron migration
neural tube closure
aortic valve morphogenesis
mitral valve morphogenesis
endocardial cushion morphogenesis
cardiac neural crest cell migration involved in outflow tract morphogenesis
transcription, DNA-templated
muscle organ development
positive regulation of gene expression
positive regulation of epithelial to mesenchymal transition
negative regulation of phosphatidylinositol 3-kinase signaling
regulation of bone mineralization
positive regulation of fatty acid beta-oxidation
negative regulation of tumor necrosis factor production
positive regulation of tumor necrosis factor production
negative regulation of histone phosphorylation
negative regulation of histone acetylation
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
negative regulation of peroxisome proliferator activated receptor signaling pathway
outer ear morphogenesis
odontogenesis
embryonic digit morphogenesis
negative regulation of apoptotic process
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of DNA damage response, signal transduction by p53 class mediator
negative regulation of osteoblast differentiation
positive regulation of angiogenesis
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
negative regulation of skeletal muscle tissue development
embryonic cranial skeleton morphogenesis
positive regulation of epithelial cell proliferation
palate development
cranial suture morphogenesis
embryonic camera-type eye formation
eyelid development in camera-type eye
cellular response to growth factor stimulus
cellular response to hypoxia
positive regulation of monocyte chemotactic protein-1 production
positive regulation of DNA-templated transcription, initiation
positive regulation of cell motility
negative regulation of oxidative phosphorylation uncoupler activity
positive regulation of transcription regulatory region DNA binding
negative regulation of cellular senescence
positive regulation of interleukin-6 secretion
negative regulation of double-strand break repair
cell proliferation involved in heart valve development
positive regulation of endocardial cushion to mesenchymal transition involved in heart valve formation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
protein domain specific binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0