Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIRREL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032123.6 | NP_115499.5 | 14, 15 | 1869 | ||
NM_199179.3 | NP_954648.3 | 13, 14 | 1719 | |||
NM_199180.3 | NP_954649.3 | 14, 15 | 2011 | |||
XM_011527362.1 | XP_011525664.1 | 15, 16 | 2071 | |||
XM_011527363.1 | XP_011525665.1 | 15, 16 | 2052 | |||
XM_011527365.1 | XP_011525667.1 | 14, 15 | 1921 | |||
XM_011527366.1 | XP_011525668.1 | 13, 14 | 2182 | |||
XM_017027348.1 | XP_016882837.1 | 13, 14 | 2473 | |||
GenBank mRNA | AB593116.1 | - | 14, 15 | 1975 | ||
AK307865.1 | - | 13, 14 | 2227 | |||
AL136654.1 | - | 14, 15 | 1995 | |||
AY305301.1 | - | 14, 15 | 1813 | |||
AY305302.1 | - | 13, 14 | 1663 | |||
AY358742.1 | - | 14, 15 | 1997 | |||
BC007312.1 | - | 5, 6 | 544 | |||
BC064925.1 | - | 17, 18 | 1934 | 6141879 |