siRNA Details
Gene Symbol: ZBED3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032367.2 | NP_115743.1 | 3 | 1327 | ||
GenBank mRNA | BC007239.1 | - | 3 | 1327 | 2906114 |

Gene Symbol: ZBED3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032367.2 | NP_115743.1 | 3 | 1327 | ||
GenBank mRNA | BC007239.1 | - | 3 | 1327 | 2906114 |