Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LLPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032338.3 | NP_115714.1 | 3 | 363 | ||
GenBank mRNA | AK057947.1 | - | 2 | 276 | ||
AK091568.1 | - | 3 | 363 | |||
BC006002.1 | - | 3 | 359 | 4247279 | ||
BC070232.1 | - | 3 | 355 | 4594274 | ||
BC107780.1 | - | 3 | 364 | 6698895 |