Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AFAP1L2 SIRNA Assays ›
Gene Symbol:
AFAP1L2
Gene Name:
actin filament associated protein 1 like 2
Gene Aliases:
CTB-1144G6.4, KIAA1914, XB130
Chromosome Location:
Chr.10: 114280730 - 114405143 on Build GRCh38
UniGene ID:
Hs.501106
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
34304
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AFAP1L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001936.2
NP_001001936.1
3
487
NM_001287824.1
NP_001274753.1
3
487
NM_032550.3
NP_115939.1
3
487
XM_005270233.4
XP_005270290.1
4
435
XM_005270237.4
XP_005270294.1
3
211
XM_005270241.4
XP_005270298.1
4
446
XM_011540286.2
XP_011538588.1
3
257
XM_011540289.1
XP_011538591.1
3
257
XM_011540290.1
XP_011538592.1
3
257
XM_017016792.1
XP_016872281.1
3
257
XM_017016793.1
XP_016872282.1
3
257
XM_017016794.1
XP_016872283.1
3
257
XM_017016795.1
XP_016872284.1
4
435
XM_017016796.1
XP_016872285.1
3
209
XM_017016797.1
XP_016872286.1
3
209
XM_017016798.1
XP_016872287.1
3
209
XM_017016799.1
XP_016872288.1
3
257
XM_017016800.1
XP_016872289.1
3
257
XM_017016801.1
XP_016872290.1
3
257
XM_017016802.1
XP_016872291.1
3
257
XM_017016803.1
XP_016872292.1
3
257
XM_017016804.1
XP_016872293.1
3
211
XM_017016805.1
XP_016872294.1
3
209
XM_017016806.1
XP_016872295.1
3
209
XM_017016807.1
XP_016872296.1
3
257
XM_017016808.1
XP_016872297.1
3
209
XM_017016809.1
XP_016872298.1
3
209
XM_017016810.1
XP_016872299.1
4
435
XM_017016811.1
XP_016872300.1
4
435
XM_017016812.1
XP_016872301.1
4
435
XM_017016813.1
XP_016872302.1
4
435
XM_017016814.1
XP_016872303.1
4
446
XM_017016815.1
XP_016872304.1
3
257
XM_017016816.1
XP_016872305.1
3
211
XM_017016817.1
XP_016872306.1
3
211
XM_017016818.1
XP_016872307.1
3
209
XM_017016819.1
XP_016872308.1
3
257
XM_017016820.1
XP_016872309.1
4
446
XM_017016821.1
XP_016872310.1
4
446
XM_017016822.1
XP_016872311.1
3
257
XM_017016823.1
XP_016872312.1
3
209
XM_017016824.1
XP_016872313.1
3
209
XM_017016825.1
XP_016872314.1
4
446
XM_017016826.1
XP_016872315.1
4
446
XM_017016827.1
XP_016872316.1
3
211
XM_017016828.1
XP_016872317.1
3
211
XM_017016830.1
XP_016872319.1
4
446
GenBank mRNA
AF474151.1
-
3
213
AK123108.1
-
3
173
AK291712.1
-
3
211
AK294969.1
-
3
210
BC024314.1
-
3
465
3900921
BC033212.1
-
3
194
4909140
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
inflammatory response
regulation of mitotic cell cycle
regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of protein tyrosine kinase activity
SH3 domain binding
protein tyrosine kinase activator activity
signaling adaptor activity
SH2 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-zgjsk:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0