Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127219.1 | NP_001120691.1 | 2 | 252 | ||
NM_032592.3 | NP_115981.1 | 2 | 548 | |||
XM_005253170.3 | XP_005253227.2 | 2 | 691 | |||
XM_011520406.2 | XP_011518708.1 | 2 | 694 | |||
XM_011520410.2 | XP_011518712.1 | 2 | 692 | |||
XM_017018421.1 | XP_016873910.1 | 2 | 695 | |||
GenBank mRNA | AK057649.1 | - | 2 | 542 | ||
AK302515.1 | - | 2 | 196 | |||
AK304074.1 | - | 2 | 485 | |||
AK307380.1 | - | 2 | 286 | |||
AY026508.1 | - | 2 | 503 | |||
BC020197.1 | - | 2 | 522 | 3632065 | ||
DA196370.1 | - | 2 | 252 | |||
DA406116.1 | - | 2 | 538 | |||
KU178811.1 | - | 1 | 108 | |||
KU178812.1 | - | 1 | 108 | |||
KU178813.1 | - | 1 | 108 |