Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321209.1 | NP_001308138.1 | 4, 5 | 369 | ||
NM_032574.3 | NP_115963.1 | 4, 5 | 509 | |||
NR_135585.1 | 4, 5 | 362 | ||||
GenBank mRNA | AF226998.1 | - | 4, 5 | 428 | ||
AV725116.1 | - | 4, 5 | 300 | |||
BC015970.1 | - | 4, 5 | 331 | 4081996 | ||
BI752326.1 | - | 4, 5 | 330 | |||
BQ276899.1 | - | 369 |