siRNA Details
Gene Symbol: ZNF503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032772.5 | NP_116161.2 | 1 | 708 | ||
NR_120651.1 | 1 | 708 | ||||
GenBank mRNA | AK225851.1 | - | 1 | 502 | ||
AK290311.1 | - | 2 | 407 | |||
BC007494.2 | - | 1 | 569 | |||
BC011625.2 | - | 1 | 481 | 4053098 | ||
BC013011.2 | - | 1 | 342 | 3604473 | ||
BC021165.2 | - | 1 | 342 | 3830632 | ||
BX105802.1 | - | 1 | 60 |

More Information
Other Species Targeted

