Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNFT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109903.1 | NP_001103373.1 | 5, 6 | 856 | ||
NM_032814.3 | NP_116203.2 | 5, 6 | 856 | |||
GenBank mRNA | AK027533.1 | - | 6, 7 | 892 | ||
BC011878.2 | - | 5, 6 | 836 | 3532898 | ||
BQ676121.1 | - | 2, 3 | 527 |