siRNA Details
Gene Symbol: ZNF503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032772.5 | NP_116161.2 | 2 | 993 | ||
GenBank mRNA | AK092897.1 | - | 1 | 112 | ||
AK127647.1 | - | 1 | 722 | |||
AK225851.1 | - | 2 | 787 | |||
AK290311.1 | - | 3 | 692 | |||
BC011625.2 | - | 2 | 766 | 4053098 | ||
BC013011.2 | - | 2 | 627 | 3604473 | ||
BC021165.2 | - | 2 | 627 | 3830632 |

More Information
Other Species Targeted

