Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPN5 SIRNA Assays ›
          Gene Symbol:
          PTPN5
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 5
          Gene Aliases:
          PTPSTEP, STEP, STEP61
          Chromosome Location:
          Chr.11: 18727928 - 18792721 on Build GRCh38
          UniGene ID:
          Hs.79092
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 34697

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPN5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039970.1 NP_001035059.1 7 937
          NM_001278236.1 NP_001265165.1 7 1771
          NM_001278238.1 NP_001265167.1 6 900
          NM_001278239.1 NP_001265168.1 6 804
          NM_006906.1 NP_008837.1 7 1033
          NM_032781.3 NP_116170.3 7 988
          XM_011520411.2 XP_011518713.1 7 972
          XM_017018434.1 XP_016873923.1 7 972
          XM_017018435.1 XP_016873924.1 7 942
          XM_017018436.1 XP_016873925.1 6 878
          XM_017018437.1 XP_016873926.1 7 896
          XM_017018438.1 XP_016873927.1 5 657
          XM_017018439.1 XP_016873928.1 6 783
          XM_017018440.1 XP_016873929.1 7 972
          XM_017018441.1 XP_016873930.1 7 972
          GenBank mRNA AK090923.1 - 7 897
          AK127312.1 - 7 1771
          AK294374.1 - 6 640
          AK294673.1 - 6 601
          AK295604.1 - 6 761
          AK316270.1 - 6 685
          AL832541.1 - 7 1033
          BC039897.1 - 6 696
          BC064807.1 - 7 797 5726473

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          response to amphetamine
          positive regulation of receptor internalization
          protein dephosphorylation
          positive regulation of neuron projection development
          positive regulation of protein import into nucleus, translocation
          peptidyl-tyrosine dephosphorylation
          exploration behavior
          response to immobilization stress
          response to drug
          negative regulation of MAP kinase activity
          positive regulation of long-term synaptic potentiation
          positive regulation of neuron death
          response to acetylsalicylate
          positive regulation of protein targeting to mitochondrion
          positive regulation of response to drug
          phosphotyrosine binding
          protein tyrosine phosphatase activity
          protein binding
          glutamate receptor binding
          mitogen-activated protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-wtt48:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline