Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNFT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109903.1 | NP_001103373.1 | 12 | 2122 | ||
NM_032814.3 | NP_116203.2 | 11 | 1639 | |||
GenBank mRNA | AK027533.1 | - | 12 | 1675 | ||
AK093164.1 | - | 8 | 1929 | |||
AK094682.1 | - | 1 | 1751 | |||
AY358585.1 | - | 1 | 1069 | |||
BC011878.2 | - | 7 | 2096 | 3532898 |