Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039374.4 | NP_001034463.4 | 6, 7 | 728 | ||
GenBank mRNA | AK090683.1 | - | 2, 3 | 304 | ||
AK092639.1 | - | 3, 4 | 986 | |||
BC137317.1 | - | 6, 7 | 703 | 9021314 | ||
BC137318.1 | - | 6, 7 | 703 | 9021315 | ||
DA235138.1 | - | 2, 3 | 258 |