Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278284.1 | NP_001265213.1 | 6 | 807 | ||
NM_001278287.1 | NP_001265216.1 | 6 | 723 | |||
NM_001278290.1 | NP_001265219.1 | 5 | 568 | |||
NM_001278291.1 | NP_001265220.1 | 4 | 437 | |||
NM_001278292.1 | NP_001265221.1 | 5 | 558 | |||
NM_001318135.1 | NP_001305064.1 | 6 | 685 | |||
NM_001318136.1 | NP_001305065.1 | 3 | 717 | |||
NM_032924.4 | NP_116313.3 | 6 | 689 | |||
XM_006716111.1 | XP_006716174.1 | 6 | 743 | |||
XM_017012582.1 | XP_016868071.1 | 6 | 757 | |||
XM_017012583.1 | XP_016868072.1 | 4 | 906 | |||
GenBank mRNA | AK000223.1 | - | 4 | 424 | ||
AK091984.1 | - | 3 | 689 | |||
AK315479.1 | - | 4 | 403 | |||
BC011887.2 | - | 6 | 613 | 3831842 | ||
BC013603.1 | - | 6 | 632 | 3865426 | ||
BC025265.1 | - | 6 | 622 | |||
BQ071498.1 | - | 6 | 519 | |||
BT006918.1 | - | 3 | 254 | |||
BX647182.1 | - | 5 | 1330 | |||
DA443888.1 | - | 4 | 423 | |||
X60153.1 | - | 4 | 362 |