Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122035.1 | 3 | 811 | |||
NR_122036.1 | 3 | 500 | ||||
GenBank mRNA | AF267857.1 | - | 3 | 729 | ||
AL516492.3 | - | 3 | 813 | |||
BC011781.2 | - | 3 | 713 | |||
BC066646.1 | - | 3 | 518 | 5749061 | ||
BC117267.1 | - | 2 | 271 | 40125818 | ||
CX164735.1 | - | 3 | 500 |